Benzyl-2-ethylhexyl phthalate ≥95.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAB2714-25GEA 0 CNY
TCIAB2714-25G
Benzyl-2-ethylhexyl phthalate ≥95.0% (通过GC)
Benzyl-2-ethylhexyl phthalate
分子式: C₂₃H₂₈O₄
分子量: 368,47 g/mol
闪点: 217°C
CAS号: 27215-22-1
EINECS: 248-335-3

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR