Benzyl isononyl phthalate (mixture of branched chain isomers)

供应商: TCI EUROPE
Warning

TCIAB2715-25GEA 0 CNY
TCIAB2715-25G
Benzyl isononyl phthalate (mixture of branched chain isomers)
Benzyl isononyl phthalate (mixture of branched chain isomers)
分子式: C₂₄H₃₀O₄
分子量: 382,50 g/mol
闪点: 228°C
CAS号: 126198-74-1

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) multi component mixture
Purity (Ester Value) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR