2,2-Bis(3-cyclohexyl-4-hydroxyphenyl)propane ≥98.0% (通过HPLC)
供应商: TCI EUROPE
|
Warning
|
TCIAB2749-25GEA
0
CNY
TCIAB2749-25G
2,2-Bis(3-cyclohexyl-4-hydroxyphenyl)propane ≥98.0% (通过HPLC)
2,2-Bis(3-cyclohexyl-4-hydroxyphenyl)propane
分子式:
C₂₇H₃₆O₂ 分子量: 392,58 g/mol |
CAS号:
57100-74-0 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...