2,2-Bis(2-hydroxy-5-biphenylyl)propane ≥98.0% (通过HPLC)

供应商: TCI EUROPE
Warning

TCIAB2750-5GEA 0 CNY
TCIAB2750-5G
2,2-Bis(2-hydroxy-5-biphenylyl)propane ≥98.0% (通过HPLC)
2,2-Bis(2-hydroxy-5-biphenylyl)propane
分子式: C₂₇H₂₄O₂
分子量: 380,49 g/mol
MDL编号: MFCD00272620
CAS号: 24038-68-4

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 115.0 to 124.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR