3,5-Bis(trifluoromethyl)-2-chloroaniline ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAB2777-25GEA 835 CNY
TCIAB2777-25G
3,5-Bis(trifluoromethyl)-2-chloroaniline ≥98.0% (通过GC)
3,5-Bis(trifluoromethyl)-2-chloroaniline
分子式: C₈H₄ClF₆N
分子量: 263,57 g/mol
MDL编号: MFCD04038716
CAS号: 201593-90-0

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR