4-Bromopyrene ≥95.0% (通过GC)

供应商: TCI EUROPE

TCIAB2807-1GEA 0 CNY
TCIAB2807-1G TCIAB2807-5G
4-Bromopyrene ≥95.0% (通过GC)
4-Bromopyrene
分子式: C₁₆H₉Br
分子量: 281,15 g/mol
MDL编号: MFCD00029250
CAS号: 1732-26-9

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 95.0 %
Melting point 150.0 to 154.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR