Benzyl-5-bromoamyl ether ≥95.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAB2819-5GEA
810
CNY
TCIAB2819-5G
Benzyl-5-bromoamyl ether ≥95.0% (通过GC)
Benzyl-5-bromoamyl ether
分子式:
C₁₂H₁₇BrO |
CAS号:
1014-93-3 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...