4-Bromo-β,β-difluorostyrene ≥98.0% (通过GC) stabilised

供应商: TCI EUROPE
Warning


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

TCIAB2856-1GEA 0 CNY
TCIAB2856-1G
4-Bromo-β,β-difluorostyrene ≥98.0% (通过GC) stabilised
4-Bromo-beta,beta-difluorostyrene
分子式: C₈H₅BrF₂
分子量: 219,03 g/mol
存储条件: Freezer
MDL编号: MFCD08276320
CAS号: 84750-93-6

现在订购

产品技术规格

Appearance Colorless to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR