3,5-Bis[3,5-bis(methoxycarbonyl)phenoxymethyl]phenol ≥98.0% (通过HPLC)
供应商: TCI EUROPE
TCIAB2865-200MGEA
0
CNY
TCIAB2865-200MG
3,5-Bis[3,5-bis(methoxycarbonyl)phenoxymethyl]phenol ≥98.0% (通过HPLC)
3,5-Bis[3,5-bis(methoxycarbonyl)phenoxymethyl]phenol
分子式:
C₂₈H₂₆O₁₁ 分子量: 538,51 g/mol |
CAS号:
186605-76-5 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 161.0 to 165.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...