Bicyclo[2.2.2]oct-5-ene-2,3-dicarboxylic anhydride ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAB3038-5GEA
1625
CNY
TCIAB3038-5G
Bicyclo[2.2.2]oct-5-ene-2,3-dicarboxylic anhydride ≥98.0% (通过GC)
Bicyclo[2.2.2]oct-5-ene-2,3-dicarboxylic anhydride
分子式:
C₁₀H₁₀O₃ |
CAS号:
6708-37-8 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Methanolysis Method) | min. 96.0 % |
Melting point | 146.0 to 150.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...