3,3'-Bis(trimethylsilyl)biphenyl-4,4'-diylbis(trifluoromethanesulphonate) ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Danger
|
TCIAB3047-1GEA
0
CNY
TCIAB3047-1G
3,3'-Bis(trimethylsilyl)biphenyl-4,4'-diylbis(trifluoromethanesulphonate) ≥98.0% (通过GC)
3,3'-Bis(trimethylsilyl)biphenyl-4,4'-diylbis(trifluoromethanesulphonate)
分子式:
C₂₀H₂₄F₆O₆S₂Si₂ 分子量: 594,69 g/mol |
CAS号:
828282-80-0 UN号: 3261 ADR: 8,II |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 115.0 to 119.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...