2-Butyl-6-phenylpyridine ≥97.0% (通过GC,滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAB3314-1GEA
1550
CNY
TCIAB3314-1G
2-Butyl-6-phenylpyridine ≥97.0% (通过GC,滴定分析)
2-Butyl-6-phenylpyridine
分子式:
C₁₅H₁₇N 分子量: 211,31 g/mol 沸点: 190°C |
CAS号:
60272-71-1 |
产品技术规格
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...