2-Butyl-3-(4-hydroxybenzoyl)benzofuran ≥98.0% (通过GC,滴定分析)

供应商: TCI EUROPE

TCIAB3339-25GEA 0 CNY
TCIAB3339-25G
2-Butyl-3-(4-hydroxybenzoyl)benzofuran ≥98.0% (通过GC,滴定分析)
2-Butyl-3-(4-hydroxybenzoyl)benzofuran
分子式: C₁₉H₁₈O₃
分子量: 294,35 g/mol
MDL编号: MFCD00040947
CAS号: 52490-15-0
EINECS: 257-959-5

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 117.0 to 121.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR