3-Bromo-4,5-dihydro-1H-benzo[b]azepin-2(3H)-one ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAB3393-5GEA 0 CNY
TCIAB3393-5G
3-Bromo-4,5-dihydro-1H-benzo[b]azepin-2(3H)-one ≥98.0% (通过GC)
3-Bromo-4,5-dihydro-1H-benzo[b]azepin-2(3H)-one
分子式: C10H10BrNO
分子量: 240,1000 g/mol
MDL编号: MFCD03085918
CAS号: 86499-96-9

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 175.0 to 179.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR