1-Amino-2-bromonaphthalene ≥98.0% (通过滴定分析)

供应商: TCI EUROPE
Warning

TCIAB3411-5GEA 0 CNY
TCIAB3411-5G
1-Amino-2-bromonaphthalene ≥98.0% (通过滴定分析)
1-Amino-2-bromonaphthalene
分子式: C₁₀H₈BrN
分子量: 222,09 g/mol
MDL编号: MFCD11870111
CAS号: 771-14-2

现在订购

产品技术规格

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 66.0 to 70.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR