1,3-双(2,6-二异丙基苯基)咪唑啉-2-亚基 ≥98.0% (通过滴定分析)

供应商: TCI EUROPE
Warning

Synonyms: SIPr

TCIAB3506-1GEA 0 CNY
TCIAB3506-1G
1,3-双(2,6-二异丙基苯基)咪唑啉-2-亚基 ≥98.0% (通过滴定分析)
1,3-双(2,6-二异丙基苯基)咪唑啉-2-亚基
分子式: C₂₇H₃₈N₂
分子量: 390,62 g/mol
存储条件: Refrigerator
MDL编号: MFCD10567379
CAS号: 258278-28-3

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR