(1S,3R)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid ≥97.0% (通过GC,滴定分析)
供应商: TCI EUROPE
TCIAB3539-1GEA
0
CNY
TCIAB3539-1G
(1S,3R)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid ≥97.0% (通过GC,滴定分析)
(1S,3R)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid
分子式:
C₁₂H₂₁NO₄ 分子量: 243,30 g/mol |
MDL编号:
MFCD06227743 CAS号: 222530-34-9 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Optical purity (LC) | min. 98.0 ee% |
Purity (Neutralization titration) | min. 97.0 % |
Specific rotation [a]20/D | 48.0 to 52.0 deg (C=1, MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...