1-Butyl-3-methylimidazoliumtrifluoro(trifluoromethyl)borate ≥98.0% (通过HPLC)
供应商: TCI EUROPE
|
Warning
|
TCIAB3542-5GEA
0
CNY
TCIAB3542-5G
1-Butyl-3-methylimidazoliumtrifluoro(trifluoromethyl)borate ≥98.0% (通过HPLC)
1-Butyl-3-methylimidazoliumtrifluoro(trifluoromethyl)borate
分子式:
C₉H₁₅BF₆N₂ 分子量: 276,03 g/mol |
CAS号:
741677-68-9 |
产品技术规格
Appearance | Light yellow to Brown clear liquid |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...