7-Bromoheptanoic acid ≥98.0% (通过滴定分析)

供应商: TCI EUROPE
Danger

TCIAB3671-5GEA 0 CNY
TCIAB3671-5G
7-Bromoheptanoic acid ≥98.0% (通过滴定分析)
7-Bromoheptanoic acid
分子式: C₇H₁₃BrO₂
分子量: 209,08 g/mol
沸点: 166°C
MDL编号: MFCD00037800
CAS号: 30515-28-7
EINECS: 250-225-5
UN号: 3261
ADR: 8,III

现在订购

产品技术规格

Appearance White to Light yellow powder to lump
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR