2-溴-3-辛基噻吩 ≥97.0% (通过GC)

供应商: TCI EUROPE

TCIAB3733-5GEA 980 CNY
TCIAB3733-5G
2-溴-3-辛基噻吩 ≥97.0% (通过GC)
2-溴-3-辛基噻吩
分子式: C₁₂H₁₉BrS
分子量: 275,2500 g/mol
沸点: 123°C
MDL编号: MFCD13193238
CAS号: 145543-83-5

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR