5-溴吲哚-2-羧酸 ≥98.0% (通过滴定分析)

供应商: TCI EUROPE
Warning

TCIAB3836-1GEA 505 CNY
TCIAB3836-1G
5-溴吲哚-2-羧酸 ≥98.0% (通过滴定分析)
5-溴吲哚-2-羧酸
分子式: C₉H₆BrNO₂
分子量: 240,06 g/mol
溶点: 280°C
存储条件: Freezer
MDL编号: MFCD00022705
CAS号: 7254-19-5

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR