4-Bromo-2,3-difluorophenetole ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAB3884-5GEA 0 CNY
TCIAB3884-5G
4-Bromo-2,3-difluorophenetole ≥98.0% (通过GC)
4-Bromo-2,3-difluorophenetole
分子式: C₈H₇BrF₂O
分子量: 237,04 g/mol
MDL编号: MFCD07368018
CAS号: 156573-09-0

现在订购

产品技术规格

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR