(S)-5-(Benzyloxy)-2-(((benzyloxy)carbonyl)amino)-5-oxopentanoic acid ≥98.0% (通过HPLC,滴定分析)
供应商: TCI EUROPE
TCIAB3902-5GEA
0
CNY
TCIAB3902-5G
(S)-5-(Benzyloxy)-2-(((benzyloxy)carbonyl)amino)-5-oxopentanoic acid ≥98.0% (通过HPLC,滴定分析)
(S)-5-(Benzyloxy)-2-(((benzyloxy)carbonyl)amino)-5-oxopentanoic acid
分子式:
C₂₀H₂₁NO₆ 分子量: 371,4 g/mol |
CAS号:
5680-86-4 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 74.0 to 78.0 deg-C |
Specific rotation [a]20/D | -7.0 to -10.0 deg (C=2.5, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...