2,6-Bis[(2R,4S,5S)-1-benzyl-4,5-diphenylimidazolidin-2-yl]pyridine ≥90.0% (通过HPLC)
供应商: TCI EUROPE
|
Warning
|
TCIAB3934-50MGEA
0
CNY
TCIAB3934-50MG
2,6-Bis[(2R,4S,5S)-1-benzyl-4,5-diphenylimidazolidin-2-yl]pyridine ≥90.0% (通过HPLC)
2,6-Bis[(2R,4S,5S)-1-benzyl-4,5-diphenylimidazolidin-2-yl]pyridine
分子式:
C₄₉H₄₅N₅ 分子量: 703,93 g/mol 存储条件: Freezer |
CAS号:
1223020-29-8 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 90.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...