2-Chloro-N,N-diethylaniline ≥98.0% (通过GC)
供应商: TCI EUROPE
TCIAC0154-10MLEA
0
CNY
TCIAC0154-10ML
2-Chloro-N,N-diethylaniline ≥98.0% (通过GC)
2-Chloro-N,N-diethylaniline
分子式:
C₁₀H₁₄ClN 分子量: 183,68 g/mol 沸点: 221°C |
MDL编号:
MFCD00059477 CAS号: 19372-80-6 |
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...