2'-Chloro-4-dimethylaminoazobenzene ≥98.0% (通过HPLC,总氮)

供应商: TCI EUROPE

TCIAC0157-25GEA 860 CNY
TCIAC0157-25G
2'-Chloro-4-dimethylaminoazobenzene ≥98.0% (通过HPLC,总氮)
2'-Chloro-4-dimethylaminoazobenzene
分子式: C₁₄H₁₄ClN₃
MDL编号: MFCD00059478
CAS号: 3010-47-7

现在订购

产品技术规格

Appearance Orange to Amber to Dark red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 108.0 to 112.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR