2-Chloroethylphosphoryl dichloride ≥97.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Danger

TCIAC0171-25GEA 0 CNY
TCIAC0171-25G
2-Chloroethylphosphoryl dichloride ≥97.0% (通过GC,滴定分析)
2-Chloroethylphosphoryl dichloride
分子式: C₂H₄Cl₃O₂P
分子量: 197,38 g/mol
沸点: 100°C
MDL编号: MFCD00013898
CAS号: 1455-05-6
UN号: 3265
ADR: 8,III

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR