2-Chloro-4-phenylphenol ≥96.0% (通过GC)

供应商: TCI EUROPE
Danger

TCIAC0263-25GEA 0 CNY
TCIAC0263-25G
2-Chloro-4-phenylphenol ≥96.0% (通过GC)
2-Chloro-4-phenylphenol
分子式: C₁₂H₉ClO
分子量: 204,65 g/mol
MDL编号: MFCD00045745
CAS号: 92-04-6
EINECS: 202-120-0

现在订购

产品技术规格

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 96.0 %
Melting point 77.0 to 80.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR