5α-胆甾烷-3β-醇 ≥95.0% (通过GC) (contains alpha-Cholestanol)

供应商: TCI EUROPE

Synonyms: 3β-羟基-5α-胆甾烷

TCIAC0317-25GEA 0 CNY
TCIAC0317-25G
5α-胆甾烷-3β-醇 ≥95.0% (通过GC) (contains alpha-Cholestanol)
5α-胆甾烷-3β-醇
分子式: C₂₇H₄₈O
分子量: 388,68 g/mol
MDL编号: MFCD00066413
CAS号: 80-97-7
EINECS: 201-315-8

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 138.0 to 143.0 deg-C
Specific rotation [a]20/D +20.0 to +28.0 deg (C=1, Dioxane)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR