Crocein scarlet 3B

供应商: TCI EUROPE

TCIAC0531-25GEA 160 CNY
TCIAC0531-25G
Crocein scarlet 3B
Crocein scarlet 3B
分子式: C₂₂H₁₆N₄O₇S₂
分子量: 556,47 g/mol
MDL编号: MFCD00003907
CAS号: 5413-75-2
EINECS: 226-502-1

现在订购

产品技术规格

Appearance Orange to Amber to Dark red powder to crystal
Lambda max. 508.0 to 512.0 nm (H2O)
Absorbance (E1%1cm) min. 490 (H2O, 508.0 to 512.0 nm)
Drying loss max. 10.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR