(2-Carbamoylphenoxy)acetic acid ≥99.0% (通过滴定分析)

供应商: TCI EUROPE

TCIAC0578-25GEA 0 CNY
TCIAC0578-25G
(2-Carbamoylphenoxy)acetic acid ≥99.0% (通过滴定分析)
(2-Carbamoylphenoxy)acetic acid
分子式: C₉H₉NO₄
分子量: 195,17 g/mol
MDL编号: MFCD00025472
CAS号: 25395-22-6
EINECS: 246-940-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 99.0 %
Melting point 216.0 to 223.0 deg-C
Solubility in hot Methanol within almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR