Amyl chloroformate ≥97.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Danger

TCIAC0587-25GEA 1550 CNY
TCIAC0587-25G
Amyl chloroformate ≥97.0% (通过GC,滴定分析)
Amyl chloroformate
分子式: C₆H₁₁ClO₂
分子量: 150,60 g/mol
沸点: 161°C
存储条件: Refrigerator
MDL编号: MFCD00058933
CAS号: 638-41-5
EINECS: 211-336-4
UN号: 3277
ADR: 6.1,II

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR