Cholesterol hydrogen phthalate ≥97.0% (通过滴定分析)

供应商: TCI EUROPE

TCIAC0692-10GEA 1725 CNY
TCIAC0692-10G
Cholesterol hydrogen phthalate ≥97.0% (通过滴定分析)
Cholesterol hydrogen phthalate
分子式: C₃₅H₅₀O₄
MDL编号: MFCD00046245
CAS号: 6732-01-0
EINECS: 229-783-9

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 97.0 %
Melting point 162.0 to 166.0 deg-C
Specific rotation [a]20/D -5.0 to -9.0 deg (C=2, CHCl3)
Solubility in Chloroform very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR