4'-Cyanobenzylidene-4-ethoxyaniline ≥98.0% (通过滴定分析)

供应商: TCI EUROPE
Warning

TCIAC0744-1GEA 0 CNY
TCIAC0744-1G
4'-Cyanobenzylidene-4-ethoxyaniline ≥98.0% (通过滴定分析)
4'-Cyanobenzylidene-4-ethoxyaniline
分子式: C₁₆H₁₄N₂O
分子量: 250,30 g/mol
MDL编号: MFCD00059585
CAS号: 34128-02-4

现在订购

产品技术规格

Appearance White to Light yellow to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Mesomorphic range 107.0 to 132.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR