Amyl-4-carboxyphenyl carbonate ≥96.0% (通过滴定分析)

供应商: TCI EUROPE

TCIAC0772-1GEA 0 CNY
TCIAC0772-1G
Amyl-4-carboxyphenyl carbonate ≥96.0% (通过滴定分析)
Amyl-4-carboxyphenyl carbonate
分子式: C₁₃H₁₆O₅
分子量: 252,27 g/mol
MDL编号: MFCD00059647
CAS号: 377085-56-8

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 96.0 %
Melting point 138.0 to 143.0 deg-C
Solubility in EtOH almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR