环戊基苯乙酸 ≥98.0% (通过滴定分析)

供应商: TCI EUROPE

TCIAC0808-10GEA 0 CNY
TCIAC0808-10G
环戊基苯乙酸 ≥98.0% (通过滴定分析)
环戊基苯乙酸
分子式: C₁₃H₁₆O₂
分子量: 204,27 g/mol
MDL编号: MFCD00001379
CAS号: 3900-93-4
EINECS: 223-454-3

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 100.0 to 103.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR