2-Chloro-1-methylpyridinium-p-toluenesulphonate ≥98.0% (通过滴定分析)

供应商: TCI EUROPE

TCIAC0906-25GEA 320 CNY
TCIAC0906-25G
2-Chloro-1-methylpyridinium-p-toluenesulphonate ≥98.0% (通过滴定分析)
2-Chloro-1-methylpyridinium-p-toluenesulphonate
分子式: C₁₃H₁₄ClNO₃S
分子量: 299,77 g/mol
存储条件: Refrigerator
MDL编号: MFCD00011985
CAS号: 7403-46-5
EINECS: 231-008-4

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 120.0 to 125.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR