α-律草烯 ≥93.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
Synonyms:
反,反,反-2,6,6,9-四甲基-1,4,8-环十一烷三烯
, α-丁子香烯
TCIAC0957-1MLEA
0
CNY
TCIAC0957-1ML
α-律草烯 ≥93.0% (通过GC)
α-律草烯
分子式:
C₁₅H₂₄ 分子量: 204,36 g/mol 沸点: 123°C 闪点: 90°C |
MDL编号:
MFCD00042689 CAS号: 6753-98-6 EINECS: 229-816-7 |
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 93.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...