3-(4-羧基苯基)丙酸 ≥98.0% (通过滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAC1023-5GEA
0
CNY
TCIAC1023-5G
3-(4-羧基苯基)丙酸 ≥98.0% (通过滴定分析)
3-(4-羧基苯基)丙酸
分子式:
C₁₀H₁₀O₄ 分子量: 194,19 g/mol |
MDL编号:
MFCD00016560 CAS号: 38628-51-2 |
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...