2-Naphthyl chloroformate ≥97.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Danger

TCIAC1115-5GEA 0 CNY
TCIAC1115-5G
2-Naphthyl chloroformate ≥97.0% (通过GC,滴定分析)
2-Naphthyl chloroformate
分子式: C₁₁H₇ClO₂
分子量: 206,63 g/mol
沸点: 112°C
MDL编号: MFCD00060059
CAS号: 7693-50-7
UN号: 3277
ADR: 6.1,II

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %
Melting point 62.0 to 65.0 deg-C
Solubility in Toluene very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR