5-Chloro-2-benzothiazolinone ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAC1137-25GEA 0 CNY
TCIAC1137-25G
5-Chloro-2-benzothiazolinone ≥98.0% (通过GC)
5-Chloro-2-benzothiazolinone
分子式: C₇H₄ClNOS
分子量: 185,63 g/mol
MDL编号: MFCD00060125
CAS号: 20600-44-6

现在订购

产品技术规格

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 234.0 to 238.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR