1-(2-Chloroethyl)pyrrolidine hydrochloride ≥98.0% (通过滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAC1202-25GEA
360
CNY
TCIAC1202-25G
1-(2-Chloroethyl)pyrrolidine hydrochloride ≥98.0% (通过滴定分析)
1-(2-Chloroethyl)pyrrolidine hydrochloride
分子式:
C₆H₁₂ClN 分子量: 170,08 g/mol 沸点: 237°C |
MDL编号:
MFCD00012718 CAS号: 7250-67-1 EINECS: 230-660-7 |
产品技术规格
Appearance | White to Green to Brown powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 171.0 to 174.0 deg-C |
Solubility in Water | within very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...