Cholesterol-2,4-dichlorobenzoate ≥98.0% (通过GC)
供应商: TCI EUROPE
TCIAC1260-25GEA
0
CNY
TCIAC1260-25G
Cholesterol-2,4-dichlorobenzoate ≥98.0% (通过GC)
Cholesterol-2,4-dichlorobenzoate
分子式:
C₃₄H₄₈Cl₂O₂ 分子量: 559,66 g/mol |
MDL编号:
MFCD00065268 CAS号: 32832-01-2 EINECS: 251-248-3 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 133.0 to 136.0 deg-C |
Specific rotation [a]20/D | -7.0 to -10.0 deg (C=2,chloroform) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...