4-Cyano-2'-nitrodiphenyl ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAC1300-1GEA 0 CNY
TCIAC1300-1G
4-Cyano-2'-nitrodiphenyl ≥98.0% (通过GC)
4-Cyano-2'-nitrodiphenyl
分子式: C₁₃H₈N₂O₂
分子量: 224,22 g/mol
MDL编号: MFCD00191622
CAS号: 75898-34-9

现在订购

产品技术规格

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 146.0 to 149.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR