(3R,5S,8aR)-(-)-六氢-3-苯基-5H-噁唑[3,2-a]吡啶-5-甲腈 ≥98.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE
Warning

Synonyms: (-)-2-氰基-6-苯基噁唑哌啶[3R-(3α,5β,8aβ)]-六氢-3-苯基-5H-噁唑并[3,2-a]吡啶-5-甲腈

TCIAC1363-1GEA 0 CNY
TCIAC1363-1G
(3R,5S,8aR)-(-)-六氢-3-苯基-5H-噁唑[3,2-a]吡啶-5-甲腈 ≥98.0% (通过HPLC,滴定分析)
(3R,5S,8aR)-(-)-六氢-3-苯基-5H-噁唑[3,2-a]吡啶-5-甲腈
分子式: C₁₄H₁₆N₂O
分子量: 228,30 g/mol
MDL编号: MFCD00142338
CAS号: 88056-92-2

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 80.0 to 84.0 deg-C
Specific rotation [a]20/D -275 to -295 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR