(1R)-(+)-樟脑内磺酰亚胺 ≥97.0% (以总氮计)

供应商: TCI EUROPE
Warning

Synonyms: (3aR)-(+)-4,5,6,7-四氢-8,8-二甲基-3H-3a,6-亚甲基-2,1-苯并异噻唑基-2,2-二氧化物(7R)-(+)-10,10-二甲基-3-硫杂-4-氮杂三环[5.2.1.01,5]癸-4-烯-3,3-二氧化物

TCIAC1391-5GEA 1025 CNY
TCIAC1391-5G
(1R)-(+)-樟脑内磺酰亚胺 ≥97.0% (以总氮计)
(1R)-(+)-樟脑内磺酰亚胺
分子式: C₁₀H₁₅NO₂S
分子量: 213,30 g/mol
MDL编号: MFCD00013315
CAS号: 107869-45-4

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Specific rotation [a]20/D +30.0 to +34.0 deg (C=2, CHCl3)
Solubility in Chloroform very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR