3-羧基-PROXYL ≥98.0% (通过HPLC,滴定分析) free radical
供应商: TCI EUROPE
|
Warning
|
Synonyms:
3-(羧基)-2,2,5,5-四甲基-1-吡咯烷基氧基
, 3-羧基-2,2,5,5-四甲基-1-吡咯烷基氧自由基
TCIAC1406-1GEA
1825
CNY
TCIAC1406-1G
3-羧基-PROXYL ≥98.0% (通过HPLC,滴定分析) free radical
3-羧基-PROXYL
分子式:
C₉H₁₆NO₃ 分子量: 186,23 g/mol 存储条件: Refrigerator |
MDL编号:
MFCD00003167 CAS号: 2154-68-9 EINECS: 218-448-2 |
产品技术规格
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...