3-Chloro-3'-methoxystilbene ≥97.0% (通过GC)

供应商: TCI EUROPE

TCIAC1593-1GEA 0 CNY
TCIAC1593-1G
3-Chloro-3'-methoxystilbene ≥97.0% (通过GC)
3-Chloro-3'-methoxystilbene
分子式: C₁₅H₁₃ClO
分子量: 244,72 g/mol
MDL编号: MFCD01861281
CAS号: 164220-45-5

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 41.0 to 44.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR