4'-Chloro-N-methylformanilide ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAC1619-25GEA 0 CNY
TCIAC1619-25G
4'-Chloro-N-methylformanilide ≥98.0% (通过GC)
4'-Chloro-N-methylformanilide
分子式: C₈H₈ClNO
分子量: 169,61 g/mol
沸点: 166°C
MDL编号: MFCD02093455
CAS号: 26772-93-0

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 51.0 to 54.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR