5-(4-Carboxyphenyl)-10,15,20-triphenylporphyrin ≥98.0% (通过HPLC)

供应商: TCI EUROPE

TCIAC1630-100MGEA 5350 CNY
TCIAC1630-100MG
5-(4-Carboxyphenyl)-10,15,20-triphenylporphyrin ≥98.0% (通过HPLC)
5-(4-Carboxyphenyl)-10,15,20-triphenylporphyrin
分子式: C₄₅H₃₀N₄O₂
MDL编号: MFCD01366156
CAS号: 95051-10-8

现在订购

产品技术规格

Appearance Dark purple powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Lambda max. 416.0 to 417.0 nm (THF)
Molar extinction coefficient min. 420000 (THF, 416.0 to 417.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR